Novus Biologicals
Manufacturer Code:NBP177757
Catalog # NBP177757
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal sequence of human Pin1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dod EC 5.2.1.8 peptidyl-prolyl cis/trans isomerase NIMA-interacting peptidylprolyl cis/trans isomerase NIMA-interacting 1 peptidyl-prolyl cis-trans isomerase NIMA-interacting 1 Peptidyl-prolyl cis-trans isomerase Pin1 PPIase Pin1 prolyl isomerase protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1 protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1 Rotamase Pin1 UBL5; Peptidyl-prolyl cis-trans isomerase NIMA-interacting 1; Peptidyl-prolyl cis-trans isomerase Pin1; PPIase Pin1; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1; Rotamase Pin1
Gene Aliases: DOD; PIN1; UBL5
UniProt ID: (Human) Q13526
Entrez Gene ID: (Human) 5300
Molecular Function: isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.