Abnova
Manufacturer Code:PAB17948
Catalog # 89-119-569
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the residues surrounding amino acid 296 of rat Pim1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Pim1.
For Research Use Only
Protein Aliases: non-specific serine/threonine protein kinase; pim-1 oncogene; proto-oncogene serine/threonine-protein kinase Pim-1; proviral integration site 1; Serine/threonine-protein kinase pim-1
Gene Aliases: Pim-1; Pim1
UniProt ID: (Rat) P26794
Entrez Gene ID: (Rat) 24649
Molecular Function: kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.