Novus Biologicals
Manufacturer Code:NBP273394AF532
Catalog # NP273394A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B8 |
Immunogen | Recombinant protein fragment corresponding to amino acids 42-278 of human Phosphoribosyl Pyrophosphate Amidotransferase (NP_002694) produced in E.coli. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Amidophosphoribosyltransferase; ATase; ATASE EC 2.4.2.14 glutamine phosphoribosylpyrophosphatate amidotransferase Glutamine phosphoribosylpyrophosphate amidotransferase glutamine PRPP amidotransferase GPATamidophosphoribosyltransferase phosphoribosyl pyrophosphate amidotransferase PRAT; glutamine phosphoribosylpyrophosphatate amidotransferase; Glutamine phosphoribosylpyrophosphate amidotransferase; glutamine PRPP amidotransferase; GPAT
Gene Aliases: ATASE; GPAT; PPAT; PRAT
UniProt ID: (Human) Q06203
Entrez Gene ID: (Human) 5471
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.