R&D Systems
Manufacturer Code:MAB8137SP
Catalog # MAB8137SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 875327 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Phospholipid Scramblase 1/PLSCR1 Met1-Pro84 Accession # O15162 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Ca(2+)-dependent phospholipid scramblase 1; Erythrocyte phospholipid scramblase; Erythrocyte phospholipid scramblase MmTRA1b MMTRA1BCa(2+)-dependent phospholipid scramblase 1 phospholipid scramblase 1 PL scramblase 1 PLSCR1; Mg(2+)-dependent nuclease; MmTRA1b; Phospholipid scramblase 1; PL scramblase 1
Gene Aliases: MMTRA1B; PLSCR1
UniProt ID: (Human) O15162
Entrez Gene ID: (Human) 5359
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.