Novus Biologicals
Manufacturer Code:NBP293007100UL
Catalog # NB027910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 21-145 of human PLA2G2D (NP_036532.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.1.4 GIID sPLA2 group IID secretory phospholipase A2 Phosphatidylcholine 2-acylhydrolase 2D phospholipase A2 group IID PLA2IID secretory phospholipase A2s Secretory-type PLA stroma-associated homolog sPLA2S SPLASHsPLA2-IID; GIID sPLA2; Group IID secretory phospholipase A2; Phosphatidylcholine 2-acylhydrolase 2D; phospholipase A2, group IID; PLA2IID; secretory phospholipase A2s; Secretory-type PLA, stroma-associated homolog
Gene Aliases: PLA2G2D; PLA2IID; sPLA2-IID; sPLA2S; SPLASH
UniProt ID: (Human) Q9UNK4
Entrez Gene ID: (Human) 26279
Molecular Function: hydrolase lipase phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.