Invitrogen
Manufacturer Code:PA526004
Catalog # PIPA526004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from human Phospholamban |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cardiac phospholamban; PLB; PLN phospholamban CMD1P PLB cardiac phospholamban CMH18
Gene Aliases: CMD1P; CMH18; PLB; Plm; PLN
UniProt ID: (Human) P26678, (Rat) P61016, (Mouse) P61014
Entrez Gene ID: (Human) 5350, (Rat) 64672, (Mouse) 18821
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.