Novus Biologicals
Manufacturer Code:NB1002462
Catalog # NB1002462
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence QPEACVIDDRSPDT corresponding to C-Terminus according to NP_001184151.1 NP_001184150.1 NP_006194.2 NP_001098101.1 NP_001184152.1 NP_001159371.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E3) cAMP-specific 3'-5'-cyclic phosphodiesterase 4D cAMP-specific phosphodiesterase PDE4D6 DKFZp686M11213 Drosophila) dunce-like phosphodiesterase E3 EC 3.1.4 FLJ97311 HSPDE4D phosphodiesterase 4D cAMP-specific phosphodiesterase 4D cAMP-specific (phosphodiesterase E3 dunce homolog; cAMP-specific 3',5'-cyclic phosphodiesterase 4D; cAMP-specific phosphodiesterase PDE4D6; DPDE3; PDE43; phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog, Drosophila); testicular tissue protein Li 136
Gene Aliases: ACRDYS2; DPDE3; HSPDE4D; PDE43; PDE4D; PDE4DN2; STRK1
UniProt ID: (Human) Q08499
Entrez Gene ID: (Human) 5144
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.