Invitrogen
Manufacturer Code:MA515202
Catalog # PIMA515202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | R.566.2 |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pThr389 of human p70 S6 kinase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 70 kDa ribosomal protein S6 kinase 1; 70 kDa ribosomal protein S6 kinase 1 P70S6K1 S6K-beta-1 p70 S6 kinase alpha p70 S6 kinase alpha 1 p70 S6 kinase alpha 2 p70 S6K-alpha p70 S6KA p70 ribosomal S6 kinase alpha p70-S6K 1 ribosomal protein S6 kinase I ribosomal protein S6 kinase beta-1 serine/threonine kinase 14 alpha serine/threonine-protein kinase 14A; CG10539-PA; CG10539-PB; Dp70s6k; dS6 kinase; p70 ribosomal S6 kinase; p70 ribosomal S6 kinase alpha; p70 S6 kinase; p70 S6 kinase 1; p70 S6 kinase alpha; p70 S6 kinase, alpha; p70 S6K-alpha; p70 S6KA; p70(S6K)-alpha; p70-S6K 1; p70S6 kinase; P70S6K; p70S6K kinase; P70S6K1; protein kinase-like 64F; Ribosomal protein S6 kinase beta-1; Ribosomal protein S6 kinase I; ribosomal protein S6 kinase, 70kD, polypeptide 1; ribosomal protein S6 kinase, 70kDa, polypeptide 1; ribosomal S6 protein kinase; RPS6-p70-protein kinase; S6 kinase; S6-kinase; S6K; S6K kinase; S6K-beta-1; S6k-PA; S6k-PB; S6K1; S6kinase; serine/threonine kinase 14 alpha; Serine/threonine-protein kinase 14A
Gene Aliases: 10539; 2610318I15Rik; 4732464A07Rik; 7-10; 7084; 70kDa; AA959758; AI256796; AI314060; CG10539; d-S6K; Dmel\CG10539; Dmel_CG10539; Dp70[s6k]; Dp70S6k; dps6k; DS6K; dS6K1; fs(3)07084; l(3)07084; p-S6K; p70 S6K; p70 S6KA; p70(S6K)-alpha; p70-alpha; p70-S6K; p70/85s6k; p70/S6K; p70[S6 kinase]; p70[S6K]; p70[S6kinase]; p70S6K; Pk64F; Pk?6; PS6K; RPS6KB1; S6 K; S6K; S6K-beta-1; S6K1; s6k11; S6k|S6K; STK14A
UniProt ID: (Human) P23443, (Mouse) Q8BSK8, (Rat) P67999
Entrez Gene ID: (Human) 6198, (Mouse) 72508, (Rat) 83840, (Fruit fly) 38654
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.