Invitrogen
Manufacturer Code:701064
Catalog # PI701064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | B2H9L2 |
Immunogen | Phosphopeptide corresponding to amino acids 384394 of human p70 ribosomal protein S6 kinase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 70 kDa ribosomal protein S6 kinase 1; 70 kDa ribosomal protein S6 kinase 1 P70S6K1 S6K-beta-1 p70 S6 kinase alpha p70 S6 kinase alpha 1 p70 S6 kinase alpha 2 p70 S6K-alpha p70 S6KA p70 ribosomal S6 kinase alpha p70-S6K 1 ribosomal protein S6 kinase I ribosomal protein S6 kinase beta-1 serine/threonine kinase 14 alpha serine/threonine-protein kinase 14A; p70 ribosomal S6 kinase alpha; p70 S6 kinase alpha; p70 S6 kinase, alpha; P70S6K1; Ribosomal protein S6 kinase beta-1; Ribosomal protein S6 kinase I; ribosomal protein S6 kinase, 70kDa, polypeptide 1; S6K-beta-1; serine/threonine kinase 14 alpha; Serine/threonine-protein kinase 14A
Gene Aliases: p70 S6KA; p70(S6K)-alpha; p70-alpha; p70-S6K; PS6K; RPS6KB1; S6K; S6K-beta-1; S6K1; STK14A
UniProt ID: (Human) P23443
Entrez Gene ID: (Human) 6198
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.