Invitrogen
Manufacturer Code:PA536863
Catalog # PIPA536863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide derived from human p47-phox around the phosphorylation site of Serine 370 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 47 kDa autosomal chronic granulomatous disease protein; 47 kDa neutrophil oxidase factor; NADPH oxidase organizer 2; NADPH oxidase subunit (47 kD); NADPH oxidase subunit (47kDa); NCF-1; NCF-47K; Neutrophil cytosol factor 1; Neutrophil cytosol factor 1 NCF-1 47 kDa autosomal chronic granulomatous disease protein 47 kDa neutrophil oxidase factor NCF-47K Neutrophil NADPH oxidase factor 1 Nox organizer 2 Nox-organizing protein 2 SH3 and PX domain-containing protein 1A p47-phox NCF1 NOXO2 SH3PXD1A; neutrophil cytosolic factor 1, (chronic granulomatous disease, autosomal 1); Neutrophil NADPH oxidase factor 1; Nox organizer 2; Nox-organizing protein 2; p47-phox; phagocyte oxidase 47 kDa; SH3 and PX domain-containing protein 1A
Gene Aliases: Ncf-1; NCF-47K; NCF1; NCF1A; NOXO2; p47-phox; p47<phox>; p47phox; SH3PXD1A
UniProt ID: (Human) P14598, (Mouse) Q09014
Entrez Gene ID: (Human) 653361, (Mouse) 17969, (Rat) 114553
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.