Invitrogen
Manufacturer Code:MA537230
Catalog # PIMA537230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | P38T180Y182-E3 |
Immunogen | A synthetic phospho-peptide corresponding to residues surrounding Thr180/Tyr182 of human phospho p38 MAPK. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 186F5S anon-sts23 BG:DS00797.3 CG7393 CG7393 gene product from transcript CG7393-RA CG7393-PA CRK1 CSAID-binding protein Csaids binding protein CSBP CSBP1 CSBP2 CSPB1 cytokine suppressive anti-inflammatory drug binding protein cytokine suppressive anti-inflammatory drug binding protein 1 cytokine suppressive anti-inflammatory drug-binding protein cytokine-supressive anti-inflammatory drug binding protein Dm p38b DmelCG7393 Dmel_CG7393 Dmp38b Dp38 D-p38 D-p38 MAPK dp38b D-p38b EC 2.7.11.24 ERK1 ERT2 ESTS:186F5S etID18993.8 EXIP fk28c03 hm:zeh1243 Hog I79_016605 MAP 14 MAP kinase 1 MAP kinase 14 MAP kinase 14A MAP kinase 2 MAP kinase MXI2 MAP kinase p38 MAP kinase p38 alpha MAP kinase p38a MAP kinase p38alpha MAP kinase p38b MAPK MAPK 14 MAPK 14A MAPK1 antibody MAPK14 mapk14a MAX-interacting protein 2 mitogen activated protein kinase 14 mitogen-activated protein kinase 14 Mitogen-activated protein kinase 14A mitogen-activated protein kinase p38 alpha Mitogen-activated protein kinase p38a Mitogen-activated protein kinase p38b MK14 Mpk34C Mxi2 OTTHUMP00000216563 OTTHUMP00000216566 p38 p38 alpha p38 alpha/Mxi2 p38 beta p38 MAP kinase p38 MAP kinase alpha p38 MAPK p38 mitogen activated protein kinase p38 mitogen-activated protein kinase p38a p38alpha p38-alpha p38alpha Exip p38b p38b MAP kinase p38beta p38b-PA p38Hog p38Kb p38MAPK Prkm14 PRKM15 reactive kinase RK RP1-179N16.5 SAPK2A stress-activated p38b MAP kinase stress-activated protein kinase 2A tRNA synthetase cofactor p38 wu:fk28c03 zeh1243 zp38a; CRK1; CSAID-binding protein; Csaids binding protein; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; Cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; MAP kinase 14; MAP kinase 2; MAP kinase MXI2; MAP kinase p38 alpha; MAPK 14; MAX-interacting protein 2; mitogen activated protein kinase 14; Mitogen-activated protein kinase 14; mitogen-activated protein kinase 14A; Mitogen-activated protein kinase p38 alpha; p38 alpha; p38 MAP kinase; p38 MAP kinase alpha; p38 MAPK; p38 mitogen activated protein kinase; p38alpha Exip; reactive kinase; SAPK2a; Stress-activated protein kinase 2a; tRNA synthetase cofactor p38
Gene Aliases: CRK1; CSBP; CSBP1; CSBP2; CSPB1; EXIP; Hog; MAPK14; MXI2; p38; p38-alpha; p38a; p38ALPHA; p38Hog; p38MAPK; PRKM14; PRKM15; RK; SAPK2A
UniProt ID: (Human) Q16539, (Mouse) P47811
Entrez Gene ID: (Human) 1432, (Rat) 81649, (Mouse) 26416
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.