Invitrogen
Manufacturer Code:MA515177
Catalog # PIMA515177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | F.52.8 |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pThr180/Tyr182 of human p38 MAPK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CG7393-PA; CRK1; CSAID-binding protein; CSAID-binding protein CSBP Csaids binding protein MAP kinase 14 MAP kinase Mxi2 MAP kinase p38 alpha MAPK 14 MAX-interacting protein 2 OTTHUMP00000216563 OTTHUMP00000216566 cytokine suppressive anti-inflammatory drug binding protein cytokine suppressive anti-inflammatory drug-binding protein mitogen-activated protein kinase p38 alpha p38 MAP kinase p38 mitogen activated protein kinase p38alpha Exip stress-activated protein kinase 2A; Csaids binding protein; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; Cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; LD24658p; MAP kinase 11; MAP kinase 14; MAP kinase 2; MAP kinase MXI2; MAP kinase p38 alpha; MAP kinase p38 beta; MAP kinase p38b; MAPK 11; MAPK 14; MAX-interacting protein 2; mitogen activated protein kinase 11; mitogen activated protein kinase 14; Mitogen-activated protein kinase 11; Mitogen-activated protein kinase 14; mitogen-activated protein kinase 14A; Mitogen-activated protein kinase p38 alpha; Mitogen-activated protein kinase p38 beta; mitogen-activated protein kinase p38-2; Mitogen-activated protein kinase p38b; p38 alpha; p38 MAP kinase; p38 MAP kinase alpha; p38 MAPK; p38 mitogen activated protein kinase; p38 mitogen-activated protein kinase; p38-2; p38alpha Exip; p38b-PA; protein kinase, mitogen activated kinase, 11, p38beta; reactive kinase; SAPK2a; SAPK2b; stress-activated p38b MAP kinase; Stress-activated protein kinase 2a; Stress-activated protein kinase 2b; stress-activated protein kinase-2; stress-activated protein kinase-2b; tRNA synthetase cofactor p38
Gene Aliases: 186F5S; anon-sts23; BG:DS00797.3; CG7393; CRK1; CSBP; CSBP1; CSBP2; CSPB1; D-p38; D-p38 MAPK; D-p38b; Dm p38b; Dmel\CG7393; Dmel_CG7393; Dmp38b; Dp38; Dp38b; ESTS:186F5S; EXIP; Hog; I79_016605; MAPK; MAPK11; MAPK14; Mpk34C; MXI2; P38; p38 beta; p38 MAPK; p38-2; p38-alpha; p38a; p38ALPHA; P38B; p38Beta; P38BETA2; p38Hog; p38Kb; p38MAPK; PRKM11; PRKM14; PRKM15; RK; SAPK2; SAPK2A; SAPK2B
UniProt ID: (Human) Q15759, (Human) Q16539, (Mouse) Q9WUI1, (Mouse) P47811, (Fruit fly) O61443
Entrez Gene ID: (Human) 5600, (Human) 1432, (Rat) 689314, (Mouse) 19094, (Rat) 81649, (Chinese hamster) 100689104, (Mouse) 26416, (Fruit fly) 34780
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.