Invitrogen
Manufacturer Code:PA5110139
Catalog # PIPA5110139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphorylated peptide around T180 & Y182 of human p38 MAPK (NP_620581.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 186F5S Adapter molecule crk anon-sts23 BG:DS00797.3 BOS_21364 c-Crk CG7393 Crk Crk1 Crk3 Crk-I CRKII Crk-II CrkIII Crk-III Crko CSAID-binding protein CSBP CSBP1 Csbp2 CSPB1 Cytokine suppressive anti-inflammatory drug-binding protein Dm p38b DmelCG7393 Dmel_CG7393 Dmp38b Dp38 D-p38 D-p38 MAPK dp38b D-p38b EC 2.7.11.24 ERK1 ERT2 ESTS:186F5S EXIP fk28c03 hm:zeh1243 Hog I79_016605 MAP 14 MAP kinase 1 MAP kinase 11 MAP kinase 14 MAP kinase 14A MAP kinase MXI2 MAP kinase p38 MAP kinase p38 alpha MAP kinase p38 beta MAP kinase p38a MAP kinase p38alpha MAP kinase p38b MAPK MAPK1 antibody MAPK11 MAPK14 mapk14a MAX-interacting protein 2 Mitogen-activated protein kinase 11 Mitogen-activated protein kinase 14 Mitogen-activated protein kinase 14A Mitogen-activated protein kinase p38 alpha Mitogen-activated protein kinase p38 beta Mitogen-activated protein kinase p38a Mitogen-activated protein kinase p38b MK14 Mpk34C MXI2 OTTHUMP00000216563 OTTHUMP00000216566 p38 p38 beta p38 MAPK p38-2 p38a p38ALPHA p38-alpha p38b p38beta P38BETA2 p38Hog p38Kb p38MAPK PRKM11 Prkm14 PRKM15 Proto-oncogene c-Crk RK RP1-179N16.5 SAPK2 SAPK2A SAPK2B Stress-activated protein kinase 2a Stress-activated protein kinase 2b wu:fk28c03 zeh1243 zp38a; CRK1; CSAID-binding protein; Csaids binding protein; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; Cytokine suppressive anti-inflammatory drug-binding protein; MAP kinase 14; MAP kinase MXI2; MAP kinase p38 alpha; MAPK 14; MAX-interacting protein 2; mitogen activated protein kinase 14; Mitogen-activated protein kinase 14; Mitogen-activated protein kinase p38 alpha; p38 alpha; p38 MAP kinase; p38 MAP kinase alpha; p38 MAPK; p38 mitogen activated protein kinase; p38alpha Exip; SAPK2a; Stress-activated protein kinase 2a; tRNA synthetase cofactor p38
Gene Aliases: Crk1; CSBP; CSBP1; CSBP2; CSPB1; EXIP; MAPK14; MXI2; p38; p38-alpha; p38a; p38ALPHA; p38MAPK; PRKM14; PRKM15; RK; SAPK2A
UniProt ID: (Human) Q16539, (Mouse) P47811
Entrez Gene ID: (Human) 1432, (Mouse) 26416
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.