Invitrogen
Manufacturer Code:MA515218
Catalog # PIMA515218
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | C.7.8 |
Immunogen | Synthetic phosphopeptide corresponding to residues around pThr180/Tyr182 of human p38 MAPK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CRK1; CSAID-binding protein; CSAID-binding protein CSBP Csaids binding protein MAP kinase 14 MAP kinase Mxi2 MAP kinase p38 alpha MAPK 14 MAX-interacting protein 2 OTTHUMP00000216563 OTTHUMP00000216566 cytokine suppressive anti-inflammatory drug binding protein cytokine suppressive anti-inflammatory drug-binding protein mitogen-activated protein kinase p38 alpha p38 MAP kinase p38 mitogen activated protein kinase p38alpha Exip stress-activated protein kinase 2A; Csaids binding protein; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; Cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; etID18993.8; MAP kinase 11; MAP kinase 14; MAP kinase 14A; MAP kinase 2; MAP kinase MXI2; MAP kinase p38 alpha; MAP kinase p38 beta; MAP kinase p38a; MAPK 11; MAPK 14; MAPK 14A; MAX-interacting protein 2; mitogen activated protein kinase 11; mitogen activated protein kinase 14; Mitogen-activated protein kinase 11; Mitogen-activated protein kinase 14; Mitogen-activated protein kinase 14A; Mitogen-activated protein kinase p38 alpha; Mitogen-activated protein kinase p38 beta; mitogen-activated protein kinase p38-2; Mitogen-activated protein kinase p38a; p38 alpha; p38 MAP kinase; p38 MAP kinase alpha; p38 MAPK; p38 mitogen activated protein kinase; p38-2; p38alpha Exip; protein kinase, mitogen activated kinase, 11, p38beta; reactive kinase; SAPK2a; SAPK2b; Stress-activated protein kinase 2a; Stress-activated protein kinase 2b; stress-activated protein kinase-2; stress-activated protein kinase-2b; tRNA synthetase cofactor p38
Gene Aliases: CRK1; CSBP; CSBP1; CSBP2; CSPB1; EXIP; fk28c03; hm:zeh1243; Hog; MAPK11; MAPK14; mapk14a; MXI2; p38; p38-2; p38-alpha; p38a; p38ALPHA; P38B; p38Beta; P38BETA2; p38Hog; p38MAPK; PRKM11; PRKM14; PRKM15; RK; SAPK2; SAPK2A; SAPK2B; wu:fk28c03; zeh1243; zgc:86905; zp38a
UniProt ID: (Human) Q15759, (Human) Q16539, (Mouse) Q9WUI1, (Mouse) P47811, (Zebrafish) Q9DGE2
Entrez Gene ID: (Human) 5600, (Human) 1432, (Rat) 689314, (Mouse) 19094, (Rat) 81649, (Mouse) 26416, (Zebrafish) 415185, (Zebrafish) 65237
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.