Invitrogen
Manufacturer Code:701057
Catalog # PI701057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | B10H8L5 |
Immunogen | Phosphopeptide corresponding to amino acids 176186 of human p38 MAPK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CSAID-binding protein; Csaids binding protein; CSBP1 CSBP2 CSPB1 EXIP Mxi2 PRKM14 PRKM15 RK SAPK2A p38 p38ALPHA Mitogen-activated protein kinase 14 Mitogen-activated protein kinase p38alpha MAP kinase p38alpha Cytokine suppressive anti-inflammatory drug binding protein CSAID binding protein CSBP MAX-interacting protein 2 MAP kinase MXI2; cytokine suppressive anti-inflammatory drug binding protein; Cytokine suppressive anti-inflammatory drug-binding protein; MAP kinase 11; MAP kinase 14; MAP kinase MXI2; MAP kinase p38 alpha; MAP kinase p38 beta; MAX-interacting protein 2; Mitogen-activated protein kinase 11; Mitogen-activated protein kinase 14; Mitogen-activated protein kinase p38 alpha; Mitogen-activated protein kinase p38 beta; mitogen-activated protein kinase p38-2; p38 MAP kinase; p38 mitogen activated protein kinase; p38-2; p38alpha Exip; SAPK2a; SAPK2b; Stress-activated protein kinase 2a; Stress-activated protein kinase 2b; stress-activated protein kinase-2; stress-activated protein kinase-2b
Gene Aliases: CSBP; CSBP1; CSBP2; CSPB1; EXIP; MAPK11; MAPK14; MXI2; p38; p38-2; p38ALPHA; P38B; p38Beta; P38BETA2; PRKM11; PRKM14; PRKM15; RK; SAPK2; SAPK2A; SAPK2B
UniProt ID: (Human) Q15759, (Human) Q16539
Entrez Gene ID: (Human) 5600, (Human) 1432
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.