Invitrogen
Manufacturer Code:25907841
Catalog # 25-907-841
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4NIT4KK |
Conjugate | PE-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MAP kinase 11; MAP kinase p38 beta; MAPK 11; mitogen activated protein kinase 11; Mitogen-activated protein kinase 11; Mitogen-activated protein kinase p38 beta; mitogen-activated protein kinase p38-2; p38-2; protein kinase, mitogen activated kinase, 11, p38beta; SAPK2b; Stress-activated protein kinase 2b; stress-activated protein kinase-2; stress-activated protein kinase-2b
Gene Aliases: MAPK11; p38-2; P38B; p38Beta; P38BETA2; PRKM11; SAPK2; SAPK2B
UniProt ID: (Human) Q15759, (Mouse) Q9WUI1
Entrez Gene ID: (Human) 5600, (Mouse) 19094
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.