Invitrogen
Manufacturer Code:PA184807
Catalog # PIPA184807
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to an amino acid sequence within p38 MAPK which includes phosphorylated Thr180 and Tyr182. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C or -20°C if preferred |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CSAID-binding protein; Csaids binding protein; cytokine suppressive anti-inflammatory drug binding protein; Cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; MAP kinase 11; MAP kinase 14; MAP kinase 2; MAP kinase MXI2; MAP kinase p38 alpha; MAP kinase p38 beta; MAPK 14; MAX-interacting protein 2; Mitogen-activated protein kinase 11; Mitogen-activated protein kinase 14; mitogen-activated protein kinase 14A; Mitogen-activated protein kinase p38 alpha; Mitogen-activated protein kinase p38 beta; mitogen-activated protein kinase p38-2; p38 MAP kinase; p38 mitogen activated protein kinase; p38-2; p38alpha Exip; reactive kinase; RP1-179N16.5 CSBP1 CSBP2 CSPB1 EXIP Mxi2 PRKM14 PRKM15 RK SAPK2A p38 p38ALPHA; SAPK2a; SAPK2b; Stress-activated protein kinase 2a; Stress-activated protein kinase 2b; stress-activated protein kinase-2; stress-activated protein kinase-2b
Gene Aliases: CRK1; CSBP; CSBP1; CSBP2; CSPB1; EXIP; Hog; MAPK11; MAPK14; MXI2; p38; p38-2; p38ALPHA; P38B; p38Beta; P38BETA2; p38Hog; PRKM11; PRKM14; PRKM15; RK; SAPK2; SAPK2A; SAPK2B
UniProt ID: (Human) Q15759, (Human) Q16539
Entrez Gene ID: (Human) 5600, (Human) 1432, (Rat) 689314, (Rat) 81649
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.