Invitrogen
Manufacturer Code:PA512652
Catalog # PIPA512652
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Dot blot (DB) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S89 of human EP300 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E1A-associated protein p300; E1A-binding protein, 300kD; Histone acetyltransferase p300; Histone acetyltransferase p300 E1A-associated protein p300 EP300 P300 KAT3B; Histone butyryltransferase p300; Histone crotonyltransferase p300; p300 HAT; Protein 2-hydroxyisobutyryltransferase p300; Protein lactyltransferas p300; Protein propionyltransferase p300
Gene Aliases: EP300; KAT3B; P300; RSTS2
UniProt ID: (Human) Q09472
Entrez Gene ID: (Human) 2033
Molecular Function:
DNA binding protein
acetyltransferase
chromatin/chromatin-binding protein
nucleic acid binding
transcription cofactor
transcription factor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.