Invitrogen
Manufacturer Code:MA537145
Catalog # PIMA537145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | mTORS2448-E11 |
Immunogen | A synthetic phospho-peptide corresponding to residues surrounding Ser2448 of human phospho mTOR. |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 2-8°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 2610315D21Rik AI327068 angiopoietin-like factor CDT6 FK506 binding protein 12-rapamycin associated protein 1 FK506 binding protein 12-rapamycin associated protein 2 FK506-binding protein 12-rapamycin complex-associated protein 1 FKBP12-rapamycin complex-associated protein FKBP12-rapamycin complex-associated protein 1 FKBP-rapamycin associated protein FKBP-rapamycin associated protein (FRAP) FKBP-rapamycin-associated protein FRAP flat FRAP Frap1 FRAP2 Mammalian target of rapamycin mechanistic target of rapamycin mechanistic target of rapamycin (serine/threonine kinase) mechanistic target of rapamycin kinase MTOR m-TOR mTORC1 RAFT1 Rapamycin and FKBP12 target 1 rapamycin and FKBP12 target-1 protein rapamycin associated protein FRAP2 rapamycin target protein 1 RAPT1 serine/threonine-protein kinase mTOR SKS; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; Mammalian target of rapamycin; Mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mTOR; Rapamycin and FKBP12 target 1; rapamycin associated protein FRAP2; Rapamycin target protein 1; RAPT1; Serine/threonine-protein kinase mTOR
Gene Aliases: 2610315D21Rik; AI327068; flat; FRAP; FRAP1; FRAP2; MTOR; RAFT1; RAPT1; SKS
UniProt ID: (Human) P42345, (Mouse) Q9JLN9
Entrez Gene ID: (Human) 2475, (Mouse) 56717
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
nucleic acid binding
nucleotide kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.