Invitrogen
Manufacturer Code:PA535397
Catalog # PIPA535397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser862 of rat GRM7 conjugated to KLH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor GRM7; G protein-coupled receptor, family C, group 1, member G; GRM7 GLUR7 MGLU7 MGLUR7 GPRC1G; Metabotropic glutamate receptor 7; metabotropic glutamate receptor mGluR7; metabotropic glutamate receptor subtype 7b; mGlu7a receptor; mGluR7
Gene Aliases: 6330570A01Rik; BB176677; C030018L03; E130018M02Rik; Gpr1g; Gprc1g; Grm7; Mglur7
UniProt ID: (Rat) P35400, (Mouse) Q68ED2
Entrez Gene ID: (Rat) 81672, (Mouse) 108073
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.