Invitrogen
Manufacturer Code:PA599478
Catalog # PIPA599478
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human hnRPD around the phosphorylation site of Ser83. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARE-binding protein AUFI type A AUF1 AUF1A AU-rich element RNA binding protein 1 37kDa AU-rich element RNA-binding factor 1 AU-rich element RNA-binding protein 1 heterogeneous nuclear ribonucleoprotein D heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA binding protein 1 37kDa) heterogeneous nuclear ribonucleoprotein D0 hnRNP D0 Hnrnpd hnRNPD0 Hnrpd P37 RNA binding protein p45AUF1; ARE-binding protein AUFI, type A; AU-rich element RNA binding protein 1, 37kDa; AU-rich element RNA-binding protein 1; heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA binding protein 1, 37kDa); Heterogeneous nuclear ribonucleoprotein D0; hnRNP D0
Gene Aliases: AUF1; AUF1A; HNRNPD; hnRNPD0; HNRPD; P37
UniProt ID: (Human) Q14103, (Mouse) Q60668
Entrez Gene ID: (Human) 3184, (Mouse) 11991
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.