Invitrogen
Manufacturer Code:PA5104859
Catalog # PIPA5104859
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human eNOS around the phosphorylation site of Thr495. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310065A03Rik cNOS constitutive NOS Ecnos EC-NOS endothelial nitric oxide synthase endothelial nitric oxide synthase 3 endothelial nitric oxide synthase NOS3 endothelial NOS eNOS e-NOS nitric oxide synthase nitric oxide synthase 3 nitric oxide synthase 3 (endothelial cell) nitric oxide synthase 3 transcript variant eNOS-delta20 nitric oxide synthase 3 transcript variant eNOS-delta20-21 nitric oxide synthase 3 transcript variant eNOS-delta21 nitric oxide synthase 3 endothelial cell nitric oxide synthase endothelial NOS NOS type III Nos3 Nos-3 NOSIII OTTHUMP00000213183; cNOS; Constitutive NOS; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; Endothelial NOS; eNOS; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; Nitric oxide synthase, endothelial; NOS type III; NOSIII
Gene Aliases: 2310065A03Rik; ECNOS; eNOS; Nos-3; NOS3
UniProt ID: (Human) P29474, (Mouse) P70313, (Rat) Q62600
Entrez Gene ID: (Human) 4846, (Mouse) 18127, (Rat) 24600
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.