Invitrogen
Manufacturer Code:PA517917
Catalog # PIPA517917
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pSer1177 of human eNOS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cNOS; Constitutive NOS; EC-NOS; EC-NOS NOS type III NOSIII OTTHUMP00000213183 cNOS constitutive NOS endothelial NOS nitric oxide synthase endothelial e-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; Endothelial NOS; eNOS; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; Nitric oxide synthase, endothelial; NOS type III; NOSIII
Gene Aliases: 2310065A03Rik; cNOS; EC-NOS; ECNOS; eNOS; NOS; Nos-3; NOS3; NOSIII
UniProt ID: (Pig) Q28969, (Human) P29474, (Mouse) P70313, (Rat) Q62600
Entrez Gene ID: (Bovine) 287024, (Pig) 397557, (Human) 4846, (Mouse) 18127, (Rat) 24600
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.