Invitrogen
Manufacturer Code:MA535847
Catalog # PIMA535847
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1532 |
Immunogen | A phospho specific peptide corresponding to residues surrounding T58/S62 of human c-Myc |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AU016757 BHLHE39 c Myc Class E basic helix-loop-helix protein 39 c-Myc mMyc MRTL Myc Myc protein Myc proto-oncogene protein Myc2 MYCC Niard Nird Proto-oncogene c-Myc RNCMYC Transcription factor p64; Avian myelocytomatosis viral (v-myc) oncogene homolog; avian myelocytomatosis viral oncogene homolog; bHLHe39; Class E basic helix-loop-helix protein 39; Myc proto-oncogene protein; myc-related translation/localization regulatory factor; myelocytomatosis viral oncogene homolog; Proto-oncogene c-Myc; Transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog
Gene Aliases: BHLHE39; c-Myc; mMyc; MRTL; MYC; MYCC; RNCMYC
UniProt ID: (Human) P01106, (Rat) P09416
Entrez Gene ID: (Human) 4609, (Rat) 24577
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.