Invitrogen
Manufacturer Code:PA5105447
Catalog # PIPA5105447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Myc around the phosphorylation site of Ser293. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU016757 Avian myelocytomatosis viral (v-myc) oncogene homolog avian myelocytomatosis viral oncogene homolog bHLHe39 c Myc class E basic helix-loop-helix protein 39 cmyc c-Myc c-myc epitope tag c-myc proto-oncogene mMyc MRTL Myc Myc protein myc proto-oncogene protein Myc2 MYCC myc-related translation/localization regulatory factor myelocytomatosis oncogene myelocytomatosis viral oncogene homolog Niard Nird proto-oncogene c-Myc RNCMYC Transcription factor p64 v-myc avian myelocytomatosis viral oncogene homolog v-myc myelocytomatosis viral oncogene homolog; Avian myelocytomatosis viral (v-myc) oncogene homolog; avian myelocytomatosis viral oncogene homolog; bHLHe39; c-myc proto-oncogene; Class E basic helix-loop-helix protein 39; Myc proto-oncogene protein; myc-related translation/localization regulatory factor; myelocytomatosis viral oncogene homolog; Proto-oncogene c-Myc; Transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog
Gene Aliases: AU016757; BHLHE39; c-Myc; mMyc; MRTL; MYC; Myc2; MYCC; Niard; Nird; RNCMYC
UniProt ID: (Human) P01106, (Rat) P09416, (Mouse) P01108
Entrez Gene ID: (Human) 4609, (Rat) 24577, (Mouse) 17869
Molecular Function: basic helix-loop-helix transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.