Invitrogen
Manufacturer Code:MA515083
Catalog # PIMA515083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | G.941.2 |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pTyr1234/1235 of human Met |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: c-Met; Hepatocyte growth factor receptor; HGF receptor; HGF receptor HGF/SF receptor SF receptor hepatocyte growth factor receptor met proto-oncogene tyrosine kinase oncogene MET proto-oncogene c-Met scatter factor receptor tyrosine-protein kinase Met; HGF/SF receptor; met proto-oncogene tyrosine kinase; Proto-oncogene c-Met; Scatter factor receptor; SF receptor; Tyrosine-protein kinase Met
Gene Aliases: AI838057; AUTS9; c-Met; DFNB97; HGF; HGFR; MET; Par4; RCCP2
UniProt ID: (Human) P08581, (Rat) P97523
Entrez Gene ID: (Human) 4233, (Rat) 24553, (Mouse) 17295
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.