Invitrogen
Manufacturer Code:PA539687
Catalog # PIPA539687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human Abl around the phosphorylation site of Tyr393/412 (D-T-YP-T-A) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abelson murine leukemia oncogene; Abelson murine leukemia viral (v-abl) oncogene homolog 1; Abelson murine leukemia viral oncogene homolog 1; Abelson tyrosine-protein kinase 1; ABL1 ABL2 ABL JTK7 p150 Proto-oncogene c-Abl Tyrosine-protein kinase ABL1 c-Abl Abelson tyrosine-protein kinase 2 Abelson murine leukemia viral oncogene homolog 2 Abelson-related gene protein Tyrosine-protein kinase ARG ABLL ARG; bcr/c-abl oncogene protein; c-abl oncogene 1, non-receptor tyrosine kinase; c-abl oncogene 1, receptor tyrosine kinase; p150; Proto-oncogene c-Abl; proto-oncogene tyrosine-protein kinase ABL1; Tyrosine-protein kinase ABL1; v-abl Abelson murine leukemia oncogene 1; v-abl Abelson murine leukemia viral oncogene 1; v-abl Abelson murine leukemia viral oncogene homolog 1
Gene Aliases: ABL; ABL1; AI325092; bcr/abl; c-ABL; c-ABL1; E430008G22Rik; JTK7; p150; v-abl
UniProt ID: (Human) P00519, (Mouse) P00520
Entrez Gene ID: (Human) 25, (Rat) 311860, (Mouse) 11350
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.