Invitrogen
Manufacturer Code:PA5114555
Catalog # PIPA5114555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human c-Abl around the phosphorylation site of Tyr245. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abelson murine leukemia oncogene; Abelson murine leukemia oncogene Abelson murine leukemia viral (v-abl) oncogene homolog 1 Abelson murine leukemia viral oncogene homolog 1 Abelson tyrosine-protein kinase 1 ABL Abl 1 antib ABL proto-oncogene 1 non-receptor tyrosine kinase Abl1 ABL2 ABLL AI325092 ARG B30 bcr/abl bcr/c-abl oncogene protein c-ABL c-abl oncogene 1 non-receptor tyrosine kinase c-abl oncogene 1 receptor tyrosine kinase c-ABL1 E430008G22Rik JTK7 p150 proto-oncogene c-Abl proto-oncogene tyrosine-protein kinase ABL1 RP11-83J21.1 Tyrosine kinase ARG Tyrosine-protein kinase ABL1 v-abl v-abl Abelson murine leukemia oncogene 1 v-abl Abelson murine leukemia viral oncogene 1 v-abl Abelson murine leukemia viral oncogene homolog 1; Abelson murine leukemia viral (v-abl) oncogene homolog 1; Abelson murine leukemia viral oncogene homolog 1; Abelson tyrosine-protein kinase 1; bcr/c-abl oncogene protein; c-abl oncogene 1, non-receptor tyrosine kinase; c-abl oncogene 1, receptor tyrosine kinase; p150; Proto-oncogene c-Abl; proto-oncogene tyrosine-protein kinase ABL1; Tyrosine-protein kinase ABL1; v-abl Abelson murine leukemia oncogene 1; v-abl Abelson murine leukemia viral oncogene 1; v-abl Abelson murine leukemia viral oncogene homolog 1
Gene Aliases: ABL; ABL1; AI325092; bcr/abl; c-ABL; c-ABL1; E430008G22Rik; JTK7; p150; v-abl
UniProt ID: (Human) P00519, (Mouse) P00520
Entrez Gene ID: (Human) 25, (Rat) 311860, (Mouse) 11350
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.