Invitrogen
Manufacturer Code:PA517915
Catalog # PIPA517915
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pSer33 pSer37 and pThr41 of human ß-catenin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: armadillo homolog; beta catenin; Beta-catenin; catenin (cadherin associated protein), beta 1, 88kDa; catenin (cadherin-associated protein), beta 1; catenin (cadherin-associated protein), beta 1, 88kDa; catenin beta; Catenin beta-1; OTTHUMP00000209289 catenin beta-1
Gene Aliases: armadillo; B-catenin; beta-catenin; Bfc; Catnb; CHBCAT; CTNNB; CTNNB1; ctnnb1-a; ctnnb1-b; id:ibd2058; Mesc; MRD19; OK/SW-cl.35; PRO2286; wu:fb73e10; wu:fi81c06; wu:fk25h01; XELAEV_18031149mg
UniProt ID: (Dog) B6V8E6, (Bovine) Q0VCX4, (Human) P35222, (Mouse) Q02248, (Rat) Q9WU82, (Zebrafish) F1QGH7
Entrez Gene ID: (Dog) 477032, (Bovine) 539003, (Chicken) 395964, (Pig) 397657, (Human) 1499, (Mouse) 12387, (Rat) 84353, (Zebrafish) 30265, (Xenopus) 399274
Molecular Function:
cell adhesion molecule
cytoskeletal protein
signaling molecule
storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.