Invitrogen
Manufacturer Code:PA5114548
Catalog # PIPA5114548
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ACTN1/2/3/4 around the phosphorylation site of Ser159. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110008F24Rik 3110023F10Rik actinin 1 smooth muscle actinin alpha 1 actinin alpha 2 actinin alpha 3 actinin alpha 3 (gene/pseudogene) actinin alpha 4 actinin alpha 1 actinin alpha 2 actinin alpha 3 ACTININ-4 ACTN1 Actn1a Actn2 Actn3 ACTN4 alpha actinin 1a alpha actinin 4 alpha-actinin cytoskeletal isoform alpha-actinin skeletal muscle Alpha-actinin skeletal muscle isoform 2 Alpha-actinin skeletal muscle isoform 3 Alpha-actinin-1 alpha-actinin-2 Alpha-actinin-3 alpha-actinin-4 BDPLT15 C77391 CMD1AA CMH23 F-actin cross-linking protein focal segmental glomerulosclerosis 1 FSGS FSGS1 non-muscle alpha-actinin 1 non-muscle alpha-actinin 4 Non-muscle alpha-actinin-1; actinin 1 smooth muscle; actinin, alpha 1; actinin, alpha 2; actinin, alpha 3; alpha actinin 1a; alpha actinin 4; Alpha-actinin cytoskeletal isoform; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 2; Alpha-actinin skeletal muscle isoform 3; Alpha-actinin-1; Alpha-actinin-2; Alpha-actinin-3; Alpha-actinin-4; F-actin cross-linking protein; focal segmental glomerulosclerosis 1; non-muscle alpha-actinin 1; Non-muscle alpha-actinin 4; Non-muscle alpha-actinin-1
Gene Aliases: 1110008F24Rik; 3110023F10Rik; ACTININ-4; ACTN1; Actn1a; ACTN2; ACTN3; ACTN4; BDPLT15; C77391; CMD1AA; CMH23; FSGS; FSGS1
UniProt ID: (Human) P12814, (Human) P35609, (Human) Q08043, (Human) O43707, (Rat) Q9Z1P2, (Mouse) Q7TPR4, (Mouse) Q9JI91, (Mouse) O88990, (Mouse) P57780, (Rat) Q9QXQ0
Entrez Gene ID: (Human) 87, (Human) 88, (Human) 89, (Human) 81, (Rat) 81634, (Mouse) 109711, (Mouse) 11472, (Rat) 291245, (Mouse) 11474, (Rat) 171009, (Mouse) 60595, (Rat) 63836
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.