Invitrogen
Manufacturer Code:MA536964
Catalog # PIMA536964
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | Zap70Y319-A3 |
Immunogen | A synthetic phospho-peptide corresponding to residues surrounding Tyr319/Tyr352 of human phospho Zap70/Syk. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 2-8°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 70 kDa zeta-associated protein; 70 kDa zeta-associated protein 70 kDa zeta-chain associated protein ADMIO2 EC 2.7.10.2 FLJ17670 FLJ17679 I79_004087 IMD48 kinase ZAP70 mrtle mur Srk STCD STD syk-related protein tyrosine kinase Syk-related tyrosine kinase Tyrosine-protein kinase ZAP-70 TZK ZA70 Zap70 ZAP-70 zeta chain of T cell receptor associated protein kinase 70 zeta chain of T cell receptor associated protein kinase 70kDa zeta-chain (TCR) associated protein kinase zeta-chain (TCR) associated protein kinase 70 zeta-chain (TCR) associated protein kinase 70kDa zeta-chain associated protein kinase 70kDa zeta-chain associated protein kinase 70kD; 70 kDa zeta-chain associated protein; Syk-related tyrosine kinase; Tyrosine-protein kinase ZAP-70; zeta chain of T cell receptor associated protein kinase 70kDa; zeta-chain (TCR) associated protein kinase 70kDa; zeta-chain associated protein kinase, 70kD
Gene Aliases: ADMIO2; IMD48; mrtle; mur; SRK; STCD; STD; TZK; ZAP-70; ZAP70
UniProt ID: (Human) P43403, (Mouse) P43404
Entrez Gene ID: (Human) 7535, (Mouse) 22637
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.