Invitrogen
Manufacturer Code:PA517815
Catalog # PIPA517815
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pTyr319 of human Zap-70 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 40 kDa tyrosine-protein kinase SYK; 70 kDa zeta-associated protein; 70 kDa zeta-associated protein syk-related tyrosine kinase tyrosine-protein kinase ZAP-70 zeta-chain associated protein kinase 70kD; 70 kDa zeta-chain associated protein; 72 kDa tyrosine-protein kinase SYK; p72-Syk; p72Syk; protein-tyrosine kinase; Spleen tyrosine kinase; syk-related protein tyrosine kinase; Syk-related tyrosine kinase; Tyrosine-protein kinase SYK; Tyrosine-protein kinase SYK-like protein; Tyrosine-protein kinase ZAP-70; zeta chain of T cell receptor associated protein kinase 70kDa; zeta-chain (TCR) associated protein kinase 70kDa; zeta-chain associated protein kinase 70kDa; zeta-chain associated protein kinase, 70kD
Gene Aliases: ADMIO2; I79_004087; IMD48; mrtle; mur; p72-Syk; p72syk; ptk72; RCJMB04_19o18; SRK; STCD; STD; SYK; Sykb; TZK; ZAP-70; ZAP70
UniProt ID: (Human) P43405, (Pig) Q00655, (Chicken) F1N9Y5, (Rat) Q64725, (Mouse) P48025, (Human) P43403, (Mouse) P43404
Entrez Gene ID: (Bovine) 515515, (Human) 6850, (Pig) 100125540, (Chicken) 427272, (Horse) 100063383, (Dog) 484196, (Rat) 25155, (Mouse) 20963, (Pig) 100739325, (Bovine) 504509, (Human) 7535, (Chicken) 420086, (Horse) 100147296, (Dog) 100271860, (Mouse) 22637, (Chinese hamster) 100767740, (Rat) 301348
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.