Invitrogen
Manufacturer Code:PA536701
Catalog # PIPA536701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide derived from human WNK1 around the phosphorylation site of Threonine 60 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Erythrocyte 65 kDa protein; Erythrocyte 65 kDa protein HSAN2 HSN2 hWNK1 KDP KIAA0344 Kinase deficient protein MGC163339 MGC163341 p65 PRKWNK1 Prostate derived sterile 20 like kinase Protein kinase lysine deficient 1 Protein kinase lysine-deficient 1 Protein kinase with no lysine 1 PSK Serine/threonine protein kinase WNK1 Serine/threonine-protein kinase WNK1 With no K WNK lysine deficient protein kinase 1 WNK1; hWNK1; Kinase deficient protein; p65; prostate-derived sterile 20-like kinase; Protein kinase lysine-deficient 1; Protein kinase with no lysine 1; protein phosphatase 1, regulatory subunit 167; Serine/threonine-protein kinase WNK1; serine/threonine-protein kinase WNK1 1; serine/threonine-protein kinase WNK1 2; WNK lysine deficient protein kinase 1 isoform
Gene Aliases: 6430573H23Rik; EG406236; HSAN2; HSN2; KDP; KIAA0344; mKIAA0344; p65; PPP1R167; PRKWNK1; PSK; WNK1
UniProt ID: (Human) Q9H4A3, (Rat) Q9JIH7, (Mouse) P83741
Entrez Gene ID: (Human) 65125, (Rat) 116477, (Mouse) 232341
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.