Invitrogen
Manufacturer Code:PA5105326
Catalog # PIPA5105326
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC32A1 around the phosphorylation site of Ser462. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA and glycine transporter; GABA and glycine transporter GVAT hVIAAT mVGAT mVIAAT R75019 rGVAT SLC32A1 solute carrier family 32 (GABA vesicular transporter) member 1 solute carrier family 32 member 1 vasicular inhibitory amino acid transporter 10D vesicular GABA transporter Vesicular inhibitory amino acid transporter VGAT VIAAT; hVIAAT; mVGAT; mVIAAT; rGVAT; solute carrier family 32 (GABA vesicular transporter), member 1; Solute carrier family 32 member 1; vasicular inhibitory amino acid transporter 10D; Vesicular GABA transporter; Vesicular inhibitory amino acid transporter
Gene Aliases: R75019; SLC32A1; VGAT; VIAAT
UniProt ID: (Human) Q9H598, (Rat) O35458, (Mouse) O35633
Entrez Gene ID: (Human) 140679, (Rat) 83612, (Mouse) 22348
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.