Invitrogen
Manufacturer Code:PA5105517
Catalog # PIPA5105517
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human VEGFR1 around the phosphorylation site of Tyr1048. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6130401C07 AI323757 CD309 Embryonic receptor kinase 2 Emrk2 Fetal liver kinase 1 fetal liver kinase-1 FLK1 FLK-1 FLK1 kinase insert domain receptor (a type III receptor tyrosine kinase) (VEGF receptor 2) FLK1 kinase insert domain receptor (VEGF receptor 2) FLT Flt1 Flt-1 fms related tyrosine kinase 1 FMS-like tyrosine kinase 1 FMS-related tyrosine kinase 1 Fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor) FRT KDR kinase insert domain protein receptor kinase insert domain receptor kinase insert domain receptor (a type III receptor tyrosine kinase) kinase NYK Krd-1 Ly73 Protein-tyrosine kinase receptor flk-1 sFlt1 soluble vascular endothelial growth factor receptor 2 soluble VEGFR 1 soluble VEGFR2 sVEGFR 1 sVEGFR-2 tyrosine kinase growth factor receptor tyrosine-protein kinase FRT Tyrosine-protein kinase receptor FLT vascular endothelial growth factor receptor 1 Vascular endothelial growth factor receptor 2 vascular endothelial growth factor receptor- 2 vascular endothelial growth factor receptor-1 vascular endothelial growth factor receptor-2 vascular endothelial growth factor receptor-3 vascular permeability factor receptor VEGF R1 VEGF receptor-2 VEGFR VEGFR 1 VEGFR1 VEGFR-1 VEGFR2 VEGFR-2; CD309; Embryonic receptor kinase 2; Fetal liver kinase 1; fetal liver kinase-1; FLK-1; FLK1 kinase insert domain receptor (a type III receptor tyrosine kinase) (VEGF receptor 2); FLK1 kinase insert domain receptor (VEGF receptor 2); FLT; FLT-1; FMS-like tyrosine kinase 1; Fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); KDR; kinase insert domain protein receptor; Kinase insert domain receptor; kinase insert domain receptor (a type III receptor tyrosine kinase); Kinase NYK; Protein-tyrosine kinase receptor flk-1; soluble vascular endothelial growth factor receptor 2; soluble VEGFR2; tyrosine kinase growth factor receptor; Tyrosine-protein kinase FRT; Tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; Vascular endothelial growth factor receptor 2; vascular endothelial growth factor receptor- 2; vascular endothelial growth factor receptor-1; vascular endothelial growth factor receptor-2; vascular endothelial growth factor receptor-3; Vascular permeability factor receptor; VEGF receptor-2; VEGFR-1; VEGFR-2
Gene Aliases: 6130401C07; AI323757; CD309; Emrk2; Flk-1; FLK1; FLT; FLT-1; FLT1; FRT; KDR; Krd-1; Ly73; sFlt1; sVEGFR-2; VEGFR; VEGFR-1; VEGFR-2; VEGFR1; VEGFR2
UniProt ID: (Human) P17948, (Mouse) P35969, (Human) P35968, (Rat) O08775, (Mouse) P35918
Entrez Gene ID: (Human) 2321, (Mouse) 14254, (Rat) 54251, (Human) 3791, (Rat) 25589, (Mouse) 16542
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.