Invitrogen
Manufacturer Code:PA5104676
Catalog # PIPA5104676
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ULK1 around the phosphorylation site of Ser467. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APG1 ATG 1 ATG1 ATG1 autophagy related 1 homolog ATG1A AU041434 Autophagy-related protein 1 homolog C. elegans EC:2.7.11.1 FLJ38455 hATG1 KIAA0722 mKIAA0722 Serine/threonine-protein kinase ULK1 serine/threonine-protein kinase Unc51.1 ULK 1 Ulk1 UNC 51 UNC51 unc-51 like autophagy activating kinase 1 unc-51 like kinase 1 Unc51.1 unc-51-like kinase 1; ATG1; ATG1 autophagy related 1 homolog; Autophagy-related protein 1 homolog; Serine/threonine-protein kinase ULK1; Serine/threonine-protein kinase Unc51.1; Unc-51-like kinase 1
Gene Aliases: ATG1; ATG1A; AU041434; hATG1; KIAA0722; mKIAA0722; ULK1; UNC51; Unc51.1
UniProt ID: (Human) O75385, (Mouse) O70405
Entrez Gene ID: (Human) 8408, (Rat) 360827, (Mouse) 22241
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.