Invitrogen
Manufacturer Code:PA538534
Catalog # PIPA538534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human Tryptophan Hydroxylase around the phosphorylation site of Ser260 (H-S-SP-D-P) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: indoleacetic acid-5-hydroxylase; L-tryptophan hydroxylase; TPRH TRPH L-tryptophan hydroxylase indoleacetic acid-5-hydroxylase tryptophan 5-hydroxylase 1 tryptophan 5-monooxygenase 1 tryptophan hydroxylase (tryptophan 5-monooxygenase); Tryptophan 5-hydroxylase 1; Tryptophan 5-monooxygenase 1; tryptophan hydroxylase (tryptophan 5-monooxygenase)
Gene Aliases: TPH; TPH1; TPRH; TRPH
UniProt ID: (Human) P17752, (Mouse) P17532, (Rat) P09810
Entrez Gene ID: (Human) 7166, (Mouse) 21990, (Rat) 24848
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.