Invitrogen
Manufacturer Code:PA5105132
Catalog # PIPA5105132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TrkA around the phosphorylation site of Tyr496. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C80751 DKFZp781I14186 EC 2.7.10.1 gp140trk high affinity nerve growth factor receptor kinase TrkA MTC neurotrophic receptor tyrosine kinase 1 Neurotrophic tyrosine kinase receptor type 1 neurotrophic tyrosine kinase receptor type 1 Ntrk1 Oncogene TRK p140-TrkA slow nerve growth factor receptor Tkr trk TRK1 TRK1-transforming tyrosine kinase protein TRKA trk-A TrkA neurotrophin receptor trkA proto-oncogene receptor tropomyosin-related kinase A Tyrosine kinase receptor Tyrosine kinase receptor A; gp140trk; High affinity nerve growth factor receptor; Neurotrophic tyrosine kinase receptor type 1; neurotrophic tyrosine kinase, receptor, type 1; Oncogene TRK; p140-TrkA; Slow nerve growth factor receptor; trk; Trk-A; TRK1-transforming tyrosine kinase protein; TrkA neurotrophin receptor; trkA proto-oncogene receptor; Tropomyosin-related kinase A; Tyrosine kinase receptor; Tyrosine kinase receptor A
Gene Aliases: C80751; MTC; NTRK1; p140-TrkA; Tkr; TRK; Trk-A; TRK1; TRKA
UniProt ID: (Human) P04629, (Mouse) Q3UFB7, (Rat) P35739
Entrez Gene ID: (Human) 4914, (Mouse) 18211, (Rat) 59109
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.