Invitrogen
Manufacturer Code:PA5105352
Catalog # PIPA5105352
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TXN around the phosphorylation site of Thr100. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADF; ADF ATL-derived factor AW550880 DKFZp686B1993 MGC61975 RP11-427L11.1 SASP surface-associated sulphydryl protein testicular tissue protein Li 199 thioredoxin thioredoxin 1 thioredoxin delta 3 TRDX TRX TRX1 Txn TXN delta 3 Txn1; ATL-derived factor; Hom s Trx; SASP; Surface-associated sulphydryl protein; testicular tissue protein Li 199; Thioredoxin; thioredoxin delta 3; Trx; TXN delta 3
Gene Aliases: ADF; AW550880; TRDX; TRX; TRX1; TXN; Txn1
UniProt ID: (Human) P10599, (Rat) P11232, (Mouse) P10639
Entrez Gene ID: (Human) 7295, (Rat) 116484, (Mouse) 22166
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.