Invitrogen
Manufacturer Code:PA5105013
Catalog # PIPA5105013
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Trk B around the phosphorylation site of Tyr516. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI848316 BDNF/NT-3 growth factors receptor BDNF-tropomyosine receptor kinase B C030027L06Rik EC 2.7.10.1 GP145 TrkB GP145TrkB GP145-TrkB GP145-TrkB/GP95-TrkB GP95 TrkB GP95-TrkB hyperphagia kinase TrkB neural receptor protein-tyrosine kinase (trkB) neurotrophic receptor tyrosine kinase 2 neurotrophic tyrosine kinase receptor type 2 neurotrophic tyrosine kinase receptor type 2 neurotrophic tyrosine receptor kinase type 2 NTRK 2 Ntrk2 OTTHUMP00000021573 OTTHUMP00000021574 OTTHUMP00000021576 OTTHUMP00000021577 RATTRKB1 Tkrb Trk B Trkb trk-B trkB protein trkB tyrosine kinase TRKB1 Tropomyosin-related kinase B tyrosine kinase receptor B; BDNF-tropomyosine receptor kinase B; BDNF/NT-3 growth factors receptor; GP145-TrkB; GP145-TrkB/GP95-TrkB; neural receptor protein-tyrosine kinase (trkB); Neurotrophic tyrosine kinase receptor type 2; neurotrophic tyrosine kinase, receptor, type 2; neurotrophic tyrosine receptor kinase type 2; Trk-B; TrkB tyrosine kinase; Tropomyosin-related kinase B; tyrosine kinase receptor B
Gene Aliases: GP145-TrkB; GP145-TrkB/GP95-TrkB; NTRK2; RATTRKB1; Tkrb; trk-B; TRKB; TRKB1
UniProt ID: (Human) Q16620, (Mouse) P15209, (Rat) Q63604
Entrez Gene ID: (Human) 4915, (Mouse) 18212, (Rat) 25054
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.