Invitrogen
Manufacturer Code:702084
Catalog # PI702084
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 6H11L46 |
Immunogen | Peptide corresponding to human Phospho-TRIM28 (Ser824) &lsqbaa819-aa827] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: E3 SUMO-protein ligase TRIM28; KAP-1; KAP1 RNF96 TIF1B; KRAB [Kruppel-associated box domain]-associated protein 1; KRAB-associated protein 1; KRAB-interacting protein 1; KRIP-1; Nuclear corepressor KAP-1; protein phosphatase 1, regulatory subunit 157; RING finger protein 96; RING-type E3 ubiquitin transferase TIF1-beta; TIF1-beta; Transcription intermediary factor 1-beta; transcriptional intermediary factor 1-beta; Tripartite motif-containing protein 28
Gene Aliases: KAP1; PPP1R157; RNF96; TF1B; TIF1B; TRIM28
UniProt ID: (Human) Q13263
Entrez Gene ID: (Human) 10155
Molecular Function:
transcription cofactor
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.