Invitrogen
Manufacturer Code:PA5105450
Catalog # PIPA5105450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TK around the phosphorylation site of Tyr181. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D530002A18Rik EC 2.7.1.21 KITH thymidine kinase 1 thymidine kinase 1 soluble isoform thymidine kinase 1 soluble thymidine kinase cytosolic thymidine kinase-1 Tk TK1 Tk-1 Tk1a Tk1b TK2; thymidine kinase 1 soluble isoform; thymidine kinase 1, soluble; Thymidine kinase, cytosolic; thymidine kinase-1
Gene Aliases: D530002A18Rik; Tk; Tk-1; TK1; Tk1a; Tk1b; TK2
UniProt ID: (Human) P04183, (Rat) P27158, (Mouse) P04184
Entrez Gene ID: (Human) 7083, (Rat) 24834, (Mouse) 21877
Molecular Function: kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.