Invitrogen
Manufacturer Code:MA537226
Catalog # PIMA537226
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | Stat6Y641-G12 |
Immunogen | A synthetic phospho-peptide corresponding to residues surrounding Tyr641 of human phospho Stat6. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 12S1644 D12S1644 IL4 STAT IL-4 Stat IL-4-STAT Signal transducer and activator of transcription 6 signal transducer and activator of transcription 6 interleukin-4 induced Signal transducer and transcription activator 6 STAT STAT interleukin4-induced Stat6 STAT-6 STAT6B STAT6C transcription factor IL-4 STAT; IL-4 Stat; Signal transducer and activator of transcription 6; signal transducer and activator of transcription 6, interleukin-4 induced; Signal transducer and transcription activator 6; STAT, interleukin4-induced; transcription factor IL-4 STAT
Gene Aliases: D12S1644; IL-4-STAT; STAT6; STAT6B; STAT6C
UniProt ID: (Human) P42226, (Mouse) P52633
Entrez Gene ID: (Human) 6778, (Mouse) 20852
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.