Invitrogen
Manufacturer Code:MA537305
Catalog # PIMA537305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | Stat1Y701-3E6 |
Immunogen | A synthetic phospho-peptide corresponding to residues surrounding Tyr701 of human phospho Stat1. |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 2-8°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 2010005J02Rik AA408197 CANDF7 DD6G4-4 DKFZp686B04100 IMD31A IMD31B IMD31C ISGF-3 OTTHUMP00000165046 OTTHUMP00000205845 Signal transducer and activator of transcription 1 signal transducer and activator of transcription 1 91kD signal transducer and activator of transcription 1 91kDa signal transducer and activator of transcription 1-alpha/beta signal transducer and activator of transcription 1-like signal transducer and activator of transcription-1 STAT STAT 1 STAT1 STAT-1 stat1 alpha STAT91 transcription factor ISGF-3 transcription factor ISGF-3 components p91/p84; signal transducer and activator of transcription 1; signal transducer and activator of transcription 1, 91kD; signal transducer and activator of transcription 1, 91kDa; Signal transducer and activator of transcription 1-alpha/beta; signal transducer and activator of transcription-1; Transcription factor ISGF-3 components p91/p84
Gene Aliases: 2010005J02Rik; AA408197; CANDF7; IMD31A; IMD31B; IMD31C; ISGF-3; STAT1; STAT91
UniProt ID: (Human) P42224
Entrez Gene ID: (Human) 6772, (Mouse) 20846
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.