Invitrogen
Manufacturer Code:MA515189
Catalog # PIMA515189
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | B.776.8 |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pTyr694 of mouse Stat5a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Mammary gland factor; mammary gland factor STAT5A; signal transducer and activator of transcription 5; Signal transducer and activator of transcription 5A
Gene Aliases: AA959963; MGF; Mpf; STAT5; STAT5A
UniProt ID: (Human) P42229, (Rat) Q62771, (Mouse) P42230
Entrez Gene ID: (Human) 6776, (Rat) 24918, (Mouse) 20850
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.