Invitrogen
Manufacturer Code:MA515193
Catalog # PIMA515193
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | R.263.6 |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pTyr705 of mouse Stat3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: acute phase response factor; Acute-phase response factor; DNA-binding protein APRF; DNA-binding protein APRF acute-phase response factor signal transducer and activator of transcription 3; Signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor)
Gene Aliases: 1110034C02Rik; ADMIO; ADMIO1; APRF; AW109958; HIES; STAT3
UniProt ID: (Bovine) P61635, (Human) P40763, (Rat) P52631, (Mouse) P42227
Entrez Gene ID: (Bovine) 508541, (Human) 6774, (Rat) 25125, (Mouse) 20848
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.