Invitrogen
Manufacturer Code:PA5105146
Catalog # PIPA5105146
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SRF around the phosphorylation site of Ser103. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW049942 AW240594 c-fos serum response element-binding transcription factor MCM1 minichromosome maintenance 1 homolog OTTHUMP00000039820 RGD1559787 serum response factor serum response factor (c-fos serum response element-binding transcription factor) SRF; c-fos serum response element-binding transcription factor; minichromosome maintenance 1 homolog; Serum response factor; serum response factor (c-fos serum response element-binding transcription factor); SRF
Gene Aliases: AW049942; AW240594; MCM1; RGD1559787; SRF
UniProt ID: (Human) P11831, (Mouse) Q9JM73
Entrez Gene ID: (Human) 6722, (Mouse) 20807, (Rat) 501099
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.