Invitrogen
Manufacturer Code:PA539813
Catalog # PIPA539813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human SREBP-1 around the phosphorylation site of Ser439 (Q-S-SP-P-L) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADD1; Adipocyte determination- and differentiation-dependent factor 1; bHLHd1; Class D basic helix-loop-helix protein 1; Processed sterol regulatory element-binding protein 1; SREBP-1; Sterol regulatory element-binding protein 1; Sterol regulatory element-binding protein 1 SREBP-1 Class D basic helix-loop-helix protein 1 bHLHd1 Sterol regulatory element-binding transcription factor 1 Processed sterol regulatory element-binding protein 1 SREBF1 BHLHD1 SREBP1 nt-SREBP1; Sterol regulatory element-binding transcription factor 1; Transcription factor SREBF1
Gene Aliases: ADD-1; ADD1; BHLHD1; SREBF1; SREBP-1; SREBP-1c; SREBP1; SREBP1a
UniProt ID: (Human) P36956, (Rat) P56720
Entrez Gene ID: (Human) 6720, (Rat) 78968
Molecular Function: basic helix-loop-helix transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.