Invitrogen
Manufacturer Code:PA539812
Catalog # PIPA539812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
peptide-ELISA (pep-ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human SPHK2 around the phosphorylation site of Thr614 (P-L-TP-P-R) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SK 2; Sphingosine kinase 2; Sphingosine kinase 2 SK 2 SPK 2; sphingosine kinase type 2; SPK 2; Spk2, Sk2
Gene Aliases: C76851; SK 2; SK-2; SK2; SPHK2; SPK 2; SPK-2
UniProt ID: (Human) Q9NRA0, (Mouse) Q9JIA7
Entrez Gene ID: (Human) 56848, (Rat) 308589, (Mouse) 56632
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.