Invitrogen
Manufacturer Code:PA5104770
Catalog # PIPA5104770
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SP1 around the phosphorylation site of Thr453. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110003E12Rik AA450830 AI845540 HGNC:11205 Sp1 Sp1 transcription factor Sp1-1 Specificity protein 1 trans-acting transcription factor 1 transcription factor Sp1 TSFP1; Specificity protein 1; trans-acting transcription factor 1; Transcription factor Sp1
Gene Aliases: 1110003E12Rik; AA450830; AI845540; SP1; Sp1-1; TSFP1
UniProt ID: (Human) P08047, (Mouse) O89090, (Rat) Q8K4R0
Entrez Gene ID: (Human) 6667, (Mouse) 20683, (Rat) 24790
Molecular Function: DNA binding protein nucleic acid binding transcription cofactor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.